Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203722_g1
          See other TRPV4 GE Assays ›
          Gene Symbol:
          Trpv4
          Gene Name:
          transient receptor potential cation channel, subfamily V, member 4
          Gene Aliases:
          0610033B08Rik, OTRPC4, Trp12, VR-OAC, VRL-2, VROAC
          Chromosome Location:
          Chr.5: 114622152 - 114658435 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203722_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpv4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022017.3 NP_071300.2 11 - 12 1931 77
          GenBank mRNA AB021875.2 - 10 - 11 1858 77
          AF208026.1 - 11 - 12 1899 77
          AF279672.1 - 11 - 12 1915 77
          AJ296078.1 - 10 - 11 1850 77
          AK083223.1 - 11 - 12 1902 77
          AK158652.1 - 7 - 8 1784 77
          BC127052.1 - 11 - 12 1896 40109678 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003063.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpv4

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          response to osmotic stress
          hypotonic response
          actin filament organization
          cell-cell junction assembly
          positive regulation of cytosolic calcium ion concentration
          osmosensory signaling pathway
          negative regulation of neuron projection development
          vasopressin secretion
          positive regulation of microtubule depolymerization
          actin cytoskeleton reorganization
          cellular response to heat
          hyperosmotic salinity response
          cortical microtubule organization
          microtubule polymerization
          regulation of response to osmotic stress
          transmembrane transport
          calcium ion import
          calcium ion transmembrane transport
          cellular response to osmotic stress
          cellular hypotonic response
          nucleotide binding
          actin binding
          osmosensor activity
          protein kinase C binding
          ion channel activity
          cation channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          ATP binding
          microtubule binding
          protein kinase binding
          SH2 domain binding
          alpha-tubulin binding
          beta-tubulin binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500027285.mm.1 TC0500003063.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027285.mm.1 NM_022017.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027285.mm.1 NM_022017.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027287.mm.1 TC0500003063.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027287.mm.1 NM_022017.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027287.mm.1 NM_022017.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027284.mm.1 TC0500003063.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027284.mm.1 NM_022017.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027284.mm.1 NM_022017.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0