Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203903_g1
          See other GM19510 GE Assays ›
          Gene Symbol:
          Gm19510
          Gene Name:
          predicted gene, 19510
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 59788417 - 59794959 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203903_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm19510
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045076.1 3 - 4 1032 89
          GenBank mRNA AK035211.1 - 3 - 4 1033 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tribbles homolog 1 (Drosophila) Trib1 A530090O15Rik,
          TRB-1,
          Trb1
          -131.3kb Anti-sense Chr. 15 - 59648654 - 59657099 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4933412E24 gene 4933412E24Rik 225.2kb Sense Chr. 15 - 60014866 - 60016618 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001599.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947472 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11872495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12454673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12306402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12248588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12289386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12399077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12135283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12110828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12454670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12372805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12258380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11918241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12289387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12402074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12033137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12013156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11918243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12110835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12026585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12033143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12110823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12250419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11966519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12399101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12259794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30256373 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500012040.mm.1 TC1500001599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012040.mm.1 NR_045076.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012040.mm.1 NR_045076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012042.mm.1 TC1500001599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012042.mm.1 NR_045076.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012042.mm.1 NR_045076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012041.mm.1 TC1500001599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012041.mm.1 NR_045076.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012041.mm.1 NR_045076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012043.mm.1 TC1500001599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012043.mm.1 NR_045076.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012043.mm.1 NR_045076.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 370313 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0