Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203954_m1
          See other D930032P07RIK GE Assays ›
          Gene Symbol:
          D930032P07Rik
          Gene Name:
          RIKEN cDNA D930032P07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 28678224 - 28720027 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203954_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D930032P07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045330.1 2 - 3 221 72
          GenBank mRNA AK086500.1 - 2 - 3 222 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000424.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125709 AK086500.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42392 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146733 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125709 AK086500.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09144948 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146733 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125709 AK086500.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14488775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14496784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14488800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14207611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14313994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14332236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14313995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14278590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14205601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14471702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14235095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14428589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14431849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32048769 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900003929.mm.1 TC1900000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003929.mm.1 NR_045330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003929.mm.1 NR_045330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003930.mm.1 TC1900000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003930.mm.1 NR_045330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003930.mm.1 NR_045330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003926.mm.1 TC1900000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003926.mm.1 NR_045330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003926.mm.1 NR_045330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003927.mm.1 TC1900000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003927.mm.1 NR_045330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003927.mm.1 NR_045330.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 766523 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104570082 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-746d97c75b-2cnrv:80/100.66.135.11:80.
          git-commit: a7e96c5202fecf029dfaed736aa540acb41e6c9b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/PDP_Cmrc_Card_Notification_Changes