Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01204140_g1
          See other PITPNM2OS1 GE Assays ›
          Gene Symbol:
          Pitpnm2os1
          Gene Name:
          phosphatidylinositol transfer protein, membrane-associated 2, opposite strand 1
          Gene Aliases:
          4930568K20Rik
          Chromosome Location:
          Chr.5: 124229725 - 124237137 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01204140_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pitpnm2os1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045369.1 5 - 6 1024 73
          GenBank mRNA AK016253.1 - 5 - 6 1023 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ADP-ribosylation factor-like 6 interacting protein 4 Arl6ip4 AA408210,
          AA408365,
          Aip-4,
          Srp25
          -118.0kb Sense Chr. 5 - 124116108 - 124118195 on NCBI Build GRCm38
          Downstream M-phase phosphoprotein 9 Mphosph9 4930548D04Rik,
          9630025B04Rik,
          AW413446,
          AW547060,
          B930097C17Rik,
          C87456,
          MPP-9,
          MPP9
          13.8kb Anti-sense Chr. 5 - 124250959 - 124330759 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001440.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2353004 AK016253.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2353004 AK016253.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04697038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04697041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04282266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04377987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24060179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04648858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431992_PM_at - -
          Mouse Genome 430 2.0 Array 1431992_PM_at - -
          Mouse Genome 430 2.0 Array 1431992_at - -
          Affymetrix Mouse Clariom S Array PSR0500012916.mm.1 TC0500001440.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012916.mm.1 NR_045369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500012916.mm.1 NR_045369.1 Refseq
          Mouse Expression Set 430 1431992_at - -
          Mouse Expression Set 430 1431992_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431992_at - -
          Affymetrix HT Mouse Genome 430B Array 1431992_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617995 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0