Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01204984_m1
          See other LIPF GE Assays ›
          Gene Symbol:
          Lipf
          Gene Name:
          lipase, gastric
          Gene Aliases:
          2310051B21Rik, AV082900
          Chromosome Location:
          Chr.19: 33919441 - 33976813 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01204984_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026334.3 NP_080610.1 6 - 7 717 158
          GenBank mRNA AK009300.1 - 6 - 7 719 158
          AK009359.1 - 6 - 7 719 158
          AK009413.1 - 6 - 7 718 158
          AK009428.1 - 6 - 7 721 158
          AK009431.1 - 6 - 7 721 158
          AK009459.1 - 6 - 7 719 158
          AK009473.1 - 6 - 7 718 158
          AK009474.1 - 6 - 7 719 158
          AK009479.1 - 6 - 7 716 158
          AK009523.1 - 6 - 7 719 158
          AK009525.1 - 6 - 7 719 158
          AK009537.1 - 6 - 7 719 158
          AK009544.1 - 6 - 7 714 158
          AK009546.1 - 6 - 7 722 158
          AK009560.1 - 6 - 7 719 158
          AK009571.1 - 6 - 7 718 158
          AK009573.1 - 6 - 7 722 158
          AK009729.1 - 6 - 7 719 158
          AK009773.1 - 6 - 7 719 158
          AK009875.1 - 6 - 7 719 158
          AK009923.1 - 6 - 7 780 158
          AK009990.1 - 6 - 7 719 158
          AK010019.1 - 6 - 7 720 158
          AK010026.1 - 6 - 7 719 158
          AK010035.1 - 6 - 7 719 158
          AK010036.1 - 6 - 7 718 158
          AK010058.1 - 6 - 7 718 158
          AK010061.1 - 6 - 7 719 158
          AK010091.1 - 6 - 7 717 158
          AK010093.1 - 6 - 7 719 158
          AK010106.1 - 6 - 7 719 158
          AK010116.1 - 6 - 7 717 158
          AK010123.1 - 6 - 7 719 157
          AK010124.1 - 5 - 6 713 158
          AK010125.1 - 6 - 7 719 158
          AK010231.1 - 6 - 7 718 158
          AK010236.1 - 6 - 7 720 158
          BC061067.1 - 6 - 7 717 30301084 158

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000488.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lipf

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          lipid catabolic process
          triglyceride lipase activity
          hydrolase activity
          hydrolase activity, acting on ester bonds

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900004355.mm.1 TC1900000488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004355.mm.1 NM_026334.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004355.mm.1 NM_026334.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0