Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01205815_m1
          See other PLCXD1 GE Assays ›
          Gene Symbol:
          Plcxd1
          Gene Name:
          phosphatidylinositol-specific phospholipase C, X domain containing 1
          Gene Aliases:
          A330045H12Rik, Gm576
          Chromosome Location:
          Chr.5: 110099225 - 110105953 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01205815_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plcxd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207279.2 NP_997162.2 2 - 3 230 97
          GenBank mRNA BC039627.1 - 2 - 3 230 5038680 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001130.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plcxd1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          molecular_function
          phosphoric diester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977773 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005941 BC039627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005941 AK149435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005941 BC034116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005941 NR_027665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005941 NM_207279.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12253 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005941 BC039627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005941 AK149435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005941 BC034116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005941 NR_027665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005941 NM_207279.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977760 - -
          Whole Mouse Genome G4122A A_51_P322658 BC039627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322658 AK149435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322658 BC034116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322658 NR_027665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322658 AK039459.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04325193 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04486320 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04325190 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04486324 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04569331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04950098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04569331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04310981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04310986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04486325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04486318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04924583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04969012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04997806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04325191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04325193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04924582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04924584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04344432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04310974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04969016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04663158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04325190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04310979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04486324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10524096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524093 - -
          Affymetrix HT MG-430 PM Array Plate 1440505_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437842_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10524089 - -
          Mouse Genome 430 2.0 Array 1440505_at - -
          Mouse Genome 430 2.0 Array 1437842_at - -
          Mouse Genome 430 2.0 Array 1437842_PM_at - -
          Mouse Genome 430 2.0 Array 1440505_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 403178_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 403178_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500009613.mm.1 TC0500001130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009613.mm.1 NM_001281812.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500009613.mm.1 NM_207279.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500009613.mm.1 NM_207279.2 Refseq
          Mouse Expression Set 430 1437842_at - -
          Mouse Expression Set 430 1440505_PM_at - -
          Mouse Expression Set 430 1437842_PM_at - -
          Mouse Expression Set 430 1440505_at - -
          Affymetrix HT Mouse Genome 430B Array 1440505_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437842_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440505_at - -
          Affymetrix HT Mouse Genome 430B Array 1437842_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407203 - -
          ABI Mouse Genome Survey Microarray 676049 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3190600 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190600 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2758190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2758190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218945 - -
          mouse-6 v1.1 (Illumina) scl0403178.9_106-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190600 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0403178.9_106-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3021892 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3094228 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2758190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2758190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3021892 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3094228 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0403178.9_106-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0