Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281812.1 | NP_001268741.1 | 2 - 3 | 154 | 77 | |
NM_207279.2 | NP_997162.2 | 3 - 4 | 321 | 77 | ||
GenBank mRNA | AK039459.1 | - | 1 - 2 | 61 | 77 | |
AK149435.1 | - | 2 - 3 | 155 | 77 | ||
BC039627.1 | - | 3 - 4 | 321 | 5038680 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |