Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
TC0500001632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
NM_001168652.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
NM_001168652.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
NM_146164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
NM_146164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
NR_033148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014847.mm.1 |
NR_033148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
TC0500001632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
NM_001168652.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
NM_001168652.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
NM_146164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
NM_146164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
NR_033148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500014848.mm.1 |
NR_033148.1 |
Refseq |