Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01211034_m1
          See other GM11837 GE Assays ›
          Gene Symbol:
          Gm11837
          Gene Name:
          predicted gene 11837
          Gene Aliases:
          OTTMUSG00000004551
          Chromosome Location:
          Chr.4: 14929829 - 14953031 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01211034_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11837
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243100.1 NP_001230029.1 4 - 5 373 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000130.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964423 NM_178617.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964423 AK081951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964423 AY278200.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964423 BC067055.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21618 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05163541 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05163530 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04003126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03621407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04021169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03621401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22747759 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400000855.mm.1 TC0400000130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000855.mm.1 NM_001243100.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000855.mm.1 NM_001243100.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0