Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01211831_g1
          See other NT5DC2 GE Assays ›
          Gene Symbol:
          Nt5dc2
          Gene Name:
          5'-nucleotidase domain containing 2
          Gene Aliases:
          2510015F01Rik
          Chromosome Location:
          Chr.14: 31131058 - 31139124 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01211831_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nt5dc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027289.1 NP_081565.1 12 - 13 1073 62
          GenBank mRNA AK010966.1 - 3 - 4 335 62
          BC011230.1 - 12 - 13 1073 4207917 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000322.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          molecular_function
          5'-nucleotidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509887 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509868 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509890 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296796 NM_027289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296796 BC011230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296796 NM_027289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296796 BC011230.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147207 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152113 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296796 NM_027289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296796 BC011230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296796 NM_027289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296796 BC011230.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152113 - -
          Whole Mouse Genome G4122A A_51_P296796 NM_027289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296796 BC011230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296796 NM_027289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296796 BC011230.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11283239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11283180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11283251 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11283256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11283241 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11283246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509887 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509868 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29509872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11727875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11273890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11775911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11273877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11802702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11591386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11802703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11189859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11243857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11283244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11802706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10413720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413719 - -
          Affymetrix HT MG-430 PM Array Plate 1424882_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10413710 - -
          Mouse Genome 430 2.0 Array 1424882_a_at - -
          Mouse Genome 430 2.0 Array 1424882_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70021_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70021_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400002767.mm.1 TC1400000322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002767.mm.1 NM_027289.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002767.mm.1 NM_027289.1 Refseq
          Mouse Expression Set 430 1424882_PM_a_at - -
          Mouse Expression Set 430 1424882_a_at - -
          Murine Genome U74v2 Set 96885_PM_at - -
          Murine Genome U74v2 Set 96885_at - -
          Mouse Genome 430A 2.0 Array 1424882_a_at - -
          Mouse Genome 430A 2.0 Array 1424882_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424882_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424882_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105080047 - -
          Illumina mouse-6 v1.0 expression beadchip 105080047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660519 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2660519 - -
          Illumina Sentrix mouse-6.v1 bead array 105080047 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105080047 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2544343 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0