Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500022830.mm.1 |
TC0500002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022830.mm.1 |
NM_001122754.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022830.mm.1 |
NM_001289494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022830.mm.1 |
NM_001289495.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022830.mm.1 |
NM_013698.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022834.mm.1 |
TC0500002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022834.mm.1 |
NM_013698.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022832.mm.1 |
TC0500002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022832.mm.1 |
NM_001122754.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022832.mm.1 |
NM_001289494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022832.mm.1 |
NM_001289495.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022832.mm.1 |
NM_013698.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022838.mm.1 |
TC0500002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022838.mm.1 |
NM_001122754.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022838.mm.1 |
NM_001289494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022838.mm.1 |
NM_001289495.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022836.mm.1 |
TC0500002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022836.mm.1 |
NM_013698.3 |
Refseq |