Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01214106_m1
          See other 2210414B05RIK GE Assays ›
          Gene Symbol:
          2210414B05Rik
          Gene Name:
          RIKEN cDNA 2210414B05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 3656506 - 3663108 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01214106_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2210414B05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040643.1 3 - 4 592 82
          GenBank mRNA AK008921.1 - 3 - 4 586 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream trimethylguanosine synthase homolog (S. cerevisiae) Tgs1 D4Ertd800e,
          Ncoa6ip,
          Pimt
          -42.6kb Sense Chr. 4 - 3574879 - 3616624 on NCBI Build GRCm38
          Downstream Yamaguchi sarcoma viral (v-yes-1) oncogene homolog Lyn AA407514,
          Hck-2,
          p53Lyn,
          p56Lyn
          15.0kb Sense Chr. 4 - 3678121 - 3791612 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000014.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20817 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111971 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111971 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109121 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03588943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03864248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03534852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03499511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03534842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03422781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03301285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03756307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04025775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03352773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04067878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22693610 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400000084.mm.1 TC0400000014.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000084.mm.1 NR_040643.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000084.mm.1 NR_040643.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887616 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0