Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00016279.mm.1 |
TC0X00002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016279.mm.1 |
NM_001160403.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016279.mm.1 |
NM_001160403.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016275.mm.1 |
TC0X00002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016275.mm.1 |
NM_001160403.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016275.mm.1 |
NM_001160403.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016276.mm.1 |
TC0X00002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016276.mm.1 |
NM_001160403.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016276.mm.1 |
NM_001160403.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016277.mm.1 |
TC0X00002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016277.mm.1 |
NM_001160403.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016277.mm.1 |
NM_001160403.1 |
Refseq |