Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216391_m1
          See other PACRGL GE Assays ›
          Gene Symbol:
          Pacrgl
          Gene Name:
          PARK2 co-regulated-like
          Gene Aliases:
          4933428G09Rik
          Chromosome Location:
          Chr.5: 48372392 - 48388756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216391_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pacrgl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025755.3 NP_080031.1 8 - 9 808 83
          GenBank mRNA AK014276.1 - 8 - 9 819 83
          AK016966.1 - 8 - 9 857 83
          AK018101.1 - 8 - 9 817 83
          AK032498.1 - 7 - 8 2556 83
          BC018481.1 - 8 - 9 808 3482169 83

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pacrgl

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671539 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 AK018101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 AK032498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 BC018481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 NM_025755.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 AK014276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 AK018101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 AK032498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 BC018481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 NM_025755.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459340 AK014276.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698451 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147488 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107587 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 AK018101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 AK032498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 BC018481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 NM_025755.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 AK014276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 AK018101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 AK032498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 BC018481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 NM_025755.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459340 AK014276.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23671533 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147488 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01094333 - -
          Whole Mouse Genome G4122A A_51_P459340 AK018101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 AK032498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 BC018481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 NM_025755.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 AK014276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 AK018101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 AK032498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 BC018481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 NM_025755.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459340 AK014276.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102048 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107587 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813332 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04502713 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04502716 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813336 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671539 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23671540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23671586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04813341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04813347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04636304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23671586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04965012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10521809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521803 - -
          Affymetrix HT MG-430 PM Array Plate 1426253_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10521798 - -
          Mouse Genome 430 2.0 Array 1426253_at - -
          Mouse Genome 430 2.0 Array 1426253_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66768_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66768_PM_at - -
          Mouse Expression Set 430 1426253_PM_at - -
          Mouse Expression Set 430 1426253_at - -
          Murine Genome U74v2 Set 104447_r_at - -
          Murine Genome U74v2 Set 104447_PM_r_at - -
          Murine Genome U74v2 Set 104446_f_at - -
          Murine Genome U74v2 Set 104446_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1426253_PM_at - -
          Mouse Genome 430A 2.0 Array 1426253_at - -
          Affymetrix HT Mouse Genome 430A Array 1426253_at - -
          Affymetrix HT Mouse Genome 430A Array 1426253_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 348797 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2510524 - -
          Illumina mouse-6 v1.0 expression beadchip 6660471 - -
          Illumina mouse-6 v1.0 expression beadchip 2510524 - -
          Illumina mouse-6 v1.0 expression beadchip 102570239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236980 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234627 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247755 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234627 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236980 - -
          mouse-6 v1.1 (Illumina) scl0004162.1_2-S - -
          mouse-6 v1.1 (Illumina) scl0004196.1_6-S - -
          mouse-6 v1.1 (Illumina) scl066768.9_199-S - -
          Illumina Sentrix mouse-6.v1 bead array 6660471 - -
          Illumina Sentrix mouse-6.v1 bead array 102570239 - -
          Illumina Sentrix mouse-6.v1 bead array 2510524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004196.1_6-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066768.9_199-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660471 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102570239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2693151 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236980 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2693151 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247755 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236980 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0