Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216587_m1
          See other GM3716 GE Assays ›
          Gene Symbol:
          Gm3716
          Gene Name:
          predicted gene 3716
          Gene Aliases:
          Gm1683
          Chromosome Location:
          Chr.5: 64593863 - 64610699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216587_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm3716
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045078.1 3 - 4 411 93
          GenBank mRNA AK044377.1 - 3 - 4 396 93
          BC038278.1 - 3 - 4 390 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA682131.1 - 3 - 4 382 Chr. 5 - 64596196 - 64610682 [-] on Build GRCm38 93
          GenBankmRNA BB642314.1 - 3 - 4 393 Chr. 5 - 64596075 - 64610697 [-] on Build GRCm38 93
          GenBankmRNA BG172788.1 - 3 - 4 376 Chr. 5 - 64595949 - 64610675 [-] on Build GRCm38 92
          GenBankmRNA BX514276.1 - 3 - 4 372 Chr. 5 - 64596156 - 64610635 [-] on Build GRCm38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA C030018K13 gene C030018K13Rik -112.0kb Anti-sense Chr. 5 - 64477008 - 64483967 on NCBI Build GRCm38
          Downstream predicted gene,
          26761
          Gm26761 91.3kb Sense Chr. 5 - 64693465 - 64699408 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002470.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55910 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964955 AK044377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964955 BC038278.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05595118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04534499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04238434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04238430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04166148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04104389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04534501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04860172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04154496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04979388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04435157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04685638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04403180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04435159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04231498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04711727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04154498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04860140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23752000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1449600_PM_at - -
          Mouse Genome 430 2.0 Array 1449600_PM_at - -
          Mouse Genome 430 2.0 Array 1449600_at - -
          Affymetrix Mouse Clariom S Array PSR0500022025.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022025.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022025.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022036.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022036.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022036.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022024.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022024.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022024.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022029.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022029.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022029.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022027.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022027.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022027.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022037.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022037.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022037.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022026.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022026.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022026.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022035.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022035.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022035.mm.1 NR_045078.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022038.mm.1 TC0500002470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022038.mm.1 NR_045078.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022038.mm.1 NR_045078.1 Refseq
          Mouse Expression Set 430 1449600_PM_at - -
          Mouse Expression Set 430 1449600_at - -
          Murine Genome U74v2 Set 96492_at - -
          Murine Genome U74v2 Set 96492_PM_at - -
          Mouse Genome 430A 2.0 Array 1449600_PM_at - -
          Mouse Genome 430A 2.0 Array 1449600_at - -
          Affymetrix HT Mouse Genome 430A Array 1449600_at - -
          Affymetrix HT Mouse Genome 430A Array 1449600_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 558512 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-777976674b-tw4cl:80/100.66.134.228:80.
          git-commit: 6354fc3e62d97773410f28d6123c879be4bd0e1c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0