Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
TC0400002470.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NM_001048005.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NM_001048005.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NM_001253777.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NM_001253777.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NM_001253788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NM_001253788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NR_045549.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019948.mm.1 |
NR_045549.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
TC0400002470.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NM_001048005.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NM_001048005.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NM_001253777.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NM_001253777.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NM_001253788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NM_001253788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NR_045549.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019950.mm.1 |
NR_045549.2 |
Refseq |