Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01217765_m1
          See other CNGA1 GE Assays ›
          Gene Symbol:
          Cnga1
          Gene Name:
          cyclic nucleotide gated channel alpha 1
          Gene Aliases:
          Cncg
          Chromosome Location:
          Chr.5: 72603696 - 72642752 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01217765_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnga1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007723.2 NP_031749.2 4 - 5 537 131
          GenBank mRNA BC141261.1 - 4 - 5 306 9055873 131
          CX564336.1 - 4 - 5 518 131
          U19717.1 - 3 - 4 272 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002578.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cnga1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          visual perception
          regulation of ion transmembrane transport
          regulation of membrane potential
          response to stimulus
          regulation of cytosolic calcium ion concentration
          membrane depolarization
          transmembrane transport
          potassium ion transmembrane transport
          cation transmembrane transport
          nucleotide binding
          ion channel activity
          intracellular cAMP-activated cation channel activity
          intracellular cGMP-activated cation channel activity
          voltage-gated potassium channel activity
          cGMP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500022797.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022797.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022797.mm.1 NM_007723.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022800.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022800.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022800.mm.1 NM_007723.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022794.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022794.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022794.mm.1 NM_007723.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022796.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022796.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022796.mm.1 NM_007723.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022795.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022795.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022795.mm.1 NM_007723.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022798.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022798.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022798.mm.1 NM_007723.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022799.mm.1 TC0500002578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022799.mm.1 NM_007723.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022799.mm.1 NM_007723.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0