Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01217808_m1
          See other CTBP1 GE Assays ›
          Gene Symbol:
          Ctbp1
          Gene Name:
          C-terminal binding protein 1
          Gene Aliases:
          BARS, CtBP1-L, CtBP1-S, CtBP3/BARS, D4S115h, D5H4S115, D5H4S115E
          Chromosome Location:
          Chr.5: 33247723 - 33275004 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01217808_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198860.1 NP_001185789.1 2 - 3 418 63
          NM_001198861.1 NP_001185790.1 2 - 3 418 63
          GenBank mRNA AK133816.1 - 2 - 3 389 63
          AK149906.1 - 2 - 3 396 63
          AK160658.1 - 2 - 3 408 63
          BC013702.1 - 1 - 2 138 3155102 63
          BC042425.1 - 2 - 3 399 5005681 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002214.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK133816.1, AK160658.1, BC013702.1, BC042425.1, NM_001198860.1, NM_001198861.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          metabolic process
          cell differentiation
          positive regulation of histone deacetylation
          negative regulation of histone acetylation
          negative regulation of transcription, DNA-templated
          white fat cell differentiation
          regulation of cell cycle
          negative regulation of histone H4 acetylation
          obsolete chromatin organization involved in negative regulation of transcription
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          DNA-binding transcription factor activity
          transcription corepressor activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          protein binding
          transcription factor binding
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          acetoin dehydrogenase activity
          protein domain specific binding
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          protein homodimerization activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          3-ketoglucose-reductase activity
          NAD binding
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
          DNA-binding transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 TC0500002214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_001198859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_001198859.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_001198860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_001198860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_001198861.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_001198861.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_013502.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020189.mm.1 NM_013502.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 TC0500002214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_001198859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_001198859.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_001198860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_001198860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_001198861.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_001198861.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_013502.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020186.mm.1 NM_013502.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020188.mm.1 TC0500002214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020188.mm.1 NM_001198860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020188.mm.1 NM_001198860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020188.mm.1 NM_001198861.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020188.mm.1 NM_001198861.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 TC0500002214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_001198859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_001198859.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_001198860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_001198860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_001198861.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_001198861.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_013502.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020191.mm.1 NM_013502.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020187.mm.1 TC0500002214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020187.mm.1 NM_001198860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020187.mm.1 NM_001198860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020187.mm.1 NM_001198861.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020187.mm.1 NM_001198861.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 925926 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0