Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198859.1 | NP_001185788.1 | 5 - 6 | 945 | 67 | |
NM_001198860.1 | NP_001185789.1 | 6 - 7 | 1138 | 67 | ||
NM_001198861.1 | NP_001185790.1 | 6 - 7 | 1138 | 67 | ||
NM_013502.3 | NP_038530.1 | 5 - 6 | 945 | 67 | ||
GenBank mRNA | AB033122.1 | - | 5 - 6 | 910 | 67 | |
AJ010483.1 | - | 5 - 6 | 858 | 67 | ||
AK133816.1 | - | 6 - 7 | 1109 | 67 | ||
AK149906.1 | - | 6 - 7 | 1114 | 67 | ||
AK160658.1 | - | 6 - 7 | 1128 | 67 | ||
AK165276.1 | - | 6 - 7 | 1093 | 67 | ||
AK170133.1 | - | 4 - 5 | 652 | 67 | ||
AK171650.1 | - | 4 - 5 | 778 | 67 | ||
AK183752.1 | - | 1 - 2 | 123 | 67 | ||
AK188829.1 | - | 1 - 2 | 163 | 67 | ||
AK195486.1 | - | 1 - 2 | 163 | 67 | ||
BC013702.1 | - | 5 - 6 | 858 | 3155102 | 67 | |
BC015071.1 | - | 5 - 6 | 914 | 4192052 | 67 | |
BC042425.1 | - | 6 - 7 | 1119 | 5005681 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |