Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218209_m1
          See other NPY2R GE Assays ›
          Gene Symbol:
          Npy2r
          Gene Name:
          neuropeptide Y receptor Y2
          Gene Aliases:
          NPY2-R
          Chromosome Location:
          Chr.3: 82538383 - 82548989 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218209_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npy2r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205099.1 NP_001192028.1 1 - 2 250 86
          NM_008731.3 NP_032757.2 1 - 2 250 86
          GenBank mRNA AK018543.1 - 1 - 2 227 86
          AK049892.1 - 1 - 2 238 86
          BC104734.1 - 1 - 2 125 40047007 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002223.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK049892.1, BC104734.1, NM_001205099.1, NM_008731.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npy2r

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          positive regulation of peptide secretion
          outflow tract morphogenesis
          cardiac left ventricle morphogenesis
          signal transduction
          G protein-coupled receptor signaling pathway
          adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          nitric oxide mediated signal transduction
          chemical synaptic transmission
          positive regulation of cell-substrate adhesion
          negative regulation of nervous system process
          positive regulation of dopamine secretion
          positive regulation of smooth muscle contraction
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          secretion
          negative regulation of synaptic transmission
          negative regulation of secretion
          regulation of sensory perception of pain
          negative regulation of synaptic transmission, glutamatergic
          negative regulation of excitatory postsynaptic potential
          negative regulation of feeding behavior
          peptide YY receptor activity
          G protein-coupled receptor activity
          neuropeptide Y receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300016726.mm.1 TC0300002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016726.mm.1 NM_001205099.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016726.mm.1 NM_001205099.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016726.mm.1 NM_008731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016726.mm.1 NM_008731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016725.mm.1 TC0300002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016725.mm.1 NM_001205099.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016725.mm.1 NM_001205099.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016725.mm.1 NM_008731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016725.mm.1 NM_008731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016727.mm.1 TC0300002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016727.mm.1 NM_001205099.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016727.mm.1 NM_001205099.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016727.mm.1 NM_008731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016727.mm.1 NM_008731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016722.mm.1 TC0300002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016722.mm.1 NM_001205099.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016722.mm.1 NM_001205099.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016722.mm.1 NM_008731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016722.mm.1 NM_008731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016720.mm.1 TC0300002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016720.mm.1 NM_001205099.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016720.mm.1 NM_001205099.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016720.mm.1 NM_008731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016720.mm.1 NM_008731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016721.mm.1 TC0300002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016721.mm.1 NM_008731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016721.mm.1 NM_008731.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0