Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218586_m1
          See other GM15408 GE Assays ›
          Gene Symbol:
          Gm15408
          Gene Name:
          predicted gene 15408
          Gene Aliases:
          OTTMUSG00000021225
          Chromosome Location:
          Chr.5: 149006962 - 149012776 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218586_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15408
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040429.1 2 - 3 473 56
          GenBank mRNA AK087624.1 - 2 - 3 205 56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX529678.1 - 2 - 3 409 Chr. 5 - 149007027 - 149011697 [+] on Build GRCm38 56
          GenBankmRNA BB666023.1 - 2 - 3 202 Chr. 5 - 149007233 - 149012341 [+] on Build GRCm38 56

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 15411 Gm15411 OTTMUSG00000021229 Anti-sense Chr. 5 - 148953741 - 149015813 on NCBI Build GRCm38
          Downstream high mobility group box 1 Hmgb1 DEF,
          HMG-1,
          Hmg1,
          SBP-1,
          amphoterin,
          p30
          35.5kb Anti-sense Chr. 5 - 149047222 - 149053037 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001834.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04421492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04217242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04421491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04550871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04651752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04726097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04217250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04569856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04263874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04950239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04484359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04484354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04217249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04726096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04550868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04238953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04217246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500016866.mm.1 TC0500001834.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016866.mm.1 NR_040429.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016866.mm.1 NR_040429.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016867.mm.1 TC0500001834.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016867.mm.1 NR_040429.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016867.mm.1 NR_040429.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 784158 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101780563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240942 - -
          Illumina Sentrix mouse-6.v1 bead array 101780563 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101780563 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2581394 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline