Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218667_m1
          See other 4930412C18RIK GE Assays ›
          Gene Symbol:
          4930412C18Rik
          Gene Name:
          RIKEN cDNA 4930412C18 gene
          Gene Aliases:
          5730591J02Rik
          Chromosome Location:
          Chr.4: 9770585 - 9823411 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218667_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930412C18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030693.1 2 - 3 223 84
          GenBank mRNA AK035353.1 - 2 - 3 224 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB627648.1 - 221 Chr. - - [-] on Build 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream aspartate-beta-hydroxylase Asph 2310005F16Rik,
          3110001L23Rik,
          AI848629,
          AW261690,
          AW561901,
          BAH,
          C79816,
          cI-37
          -113.3kb Anti-sense Chr. 4 - 9449085 - 9669344 on NCBI Build GRCm38
          Downstream growth differentiation factor 6 Gdf6 BMP-13,
          BMP13,
          GDF16
          59.2kb Sense Chr. 4 - 9844372 - 9862345 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000068.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2196185 AK076677.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2196185 NR_030693.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336999 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9422 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172991 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149426 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2196185 AK076677.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2196185 NR_030693.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05152823 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05152801 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172991 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05152721 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149426 - -
          Whole Mouse Genome G4122A A_52_P499240 AK054421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499240 AK019984.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115393 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133570 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172991 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03687046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03571112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03923596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03982906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03896358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04053056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03213454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03254005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03213465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03445546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03213468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03614067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04086561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04007412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03571107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03923602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03923597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03896360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03363693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03446136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03458102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22722374 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454040_PM_at - -
          Mouse Genome 430 2.0 Array 1454040_at - -
          Mouse Genome 430 2.0 Array 1454040_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400000455.mm.1 TC0400000068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000455.mm.1 NR_030693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000455.mm.1 NR_030693.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000456.mm.1 TC0400000068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000456.mm.1 NR_030693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000456.mm.1 NR_030693.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000457.mm.1 TC0400000068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000457.mm.1 NR_030693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000457.mm.1 NR_030693.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000458.mm.1 TC0400000068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000458.mm.1 NR_030693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000458.mm.1 NR_030693.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000454.mm.1 TC0400000068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000454.mm.1 NR_030693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000454.mm.1 NR_030693.1 Refseq
          Mouse Expression Set 430 1454040_at - -
          Mouse Expression Set 430 1454040_PM_at - -
          Murine Genome U74v2 Set 129884_at - -
          Murine Genome U74v2 Set 170133_PM_at - -
          Murine Genome U74v2 Set 170133_at - -
          Murine Genome U74v2 Set 129884_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454040_at - -
          Affymetrix HT Mouse Genome 430B Array 1454040_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100070372 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0