Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400012968.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012968.mm.1 |
NM_011894.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012970.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012970.mm.1 |
NM_011894.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012969.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012969.mm.1 |
NM_011894.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012973.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012973.mm.1 |
NM_011894.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012966.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012966.mm.1 |
NM_011894.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012967.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012967.mm.1 |
NM_011894.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012972.mm.1 |
TC1400001676.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012972.mm.1 |
NM_011894.3 |
Refseq |