Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01219855_g1
          See other NCOA4 GE Assays ›
          Gene Symbol:
          Ncoa4
          Gene Name:
          nuclear receptor coactivator 4
          Gene Aliases:
          ARA70, Rfg
          Chromosome Location:
          Chr.14: 32159887 - 32179855 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01219855_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncoa4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033988.2 NP_001029160.1 2 - 3 280 69
          NM_001284319.1 NP_001271248.1 2 - 3 267 69
          NM_019744.4 NP_062718.2 2 - 3 190 69
          GenBank mRNA AF159461.1 - 2 - 3 187 69
          AK014479.1 - 2 - 3 188 69
          AK129020.1 - 2 - 3 238 69
          AK132981.1 - 69
          AK143965.1 - 2 - 3 185 69
          AK145070.1 - 2 - 3 241 69
          AK145945.1 - 2 - 3 233 69
          AK145954.1 - 2 - 3 237 69
          AK146384.1 - 2 - 3 191 69
          AK146498.1 - 2 - 3 230 69
          AK146598.1 - 2 - 3 214 69
          AK146646.1 - 2 - 3 185 69
          AK146649.1 - 2 - 3 187 69
          AK147036.1 - 2 - 3 188 69
          AK147125.1 - 2 - 3 232 69
          AK156999.1 - 2 - 3 266 69
          BC031528.1 - 2 - 3 231 4008986 69
          BC085086.1 - 2 - 3 176 6335192 69
          CF161810.1 - 2 - 3 275 69
          CJ088075.1 - 2 - 3 283 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000340.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001284319.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ncoa4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400002959.mm.1 TC1400000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002959.mm.1 NM_019744.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002950.mm.1 TC1400000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002950.mm.1 NM_001033988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002956.mm.1 TC1400000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002956.mm.1 NM_001284319.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002963.mm.1 TC1400000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002963.mm.1 NM_001033988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002963.mm.1 NM_001284319.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002963.mm.1 NM_019744.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002964.mm.1 TC1400000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002964.mm.1 NM_001033988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002964.mm.1 NM_001284319.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002964.mm.1 NM_019744.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 569470 - -
          ABI Mouse Genome Survey Microarray 833165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0