Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080388.2 | NP_001073857.1 | 4 - 5 | 977 | 127 | |
NM_001080389.2 | NP_001073858.1 | 4 - 5 | 977 | 127 | ||
NM_001080390.2 | NP_001073859.1 | 4 - 5 | 694 | 127 | ||
NM_007928.3 | NP_031954.2 | 4 - 5 | 977 | 127 | ||
GenBank mRNA | AK045329.1 | - | 4 - 5 | 551 | 127 | |
AK154864.1 | - | 4 - 5 | 694 | 127 | ||
AK172444.1 | - | 4 - 5 | 937 | 127 | ||
AK172664.1 | - | 4 - 5 | 620 | 127 | ||
AK210480.1 | - | 3 - 4 | 128 | 127 | ||
AK218887.1 | - | 3 - 4 | 128 | 127 | ||
AK220191.1 | - | 5 - 6 | 543 | 127 | ||
BC058556.1 | - | 4 - 5 | 914 | 5289799 | 127 | |
X70764.1 | - | 4 - 5 | 431 | 127 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |