Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01220472_g1
          See other DNAJC5G GE Assays ›
          Gene Symbol:
          Dnajc5g
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 5 gamma
          Gene Aliases:
          4922503E23Rik
          Chromosome Location:
          Chr.5: 31108292 - 31112526 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01220472_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc5g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177677.3 NP_808345.1 5 - 6 639 98
          GenBank mRNA AK076638.1 - 5 - 6 640 98
          BC089502.1 - 5 - 6 618 6774833 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000312.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajc5g

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P15522 BC089502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P15522 NM_177677.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P15522 AK076638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P15522 BC089502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P15522 NM_177677.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P15522 AK076638.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165315 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P15522 BC089502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P15522 NM_177677.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P15522 AK076638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P15522 BC089502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P15522 NM_177677.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P15522 AK076638.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05522009 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05522021 - -
          Whole Mouse Genome G4122A A_51_P293496 BC089502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293496 NM_177677.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293496 AK076638.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P15522 BC089502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P15522 NM_177677.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P15522 AK076638.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P15522 BC089502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P15522 NM_177677.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P15522 AK076638.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135405 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05522021 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04401004 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04400998 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04401000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04401011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04401009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04853122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04170357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04853121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04457480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04576239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04281967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04133554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04457474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04576242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04400999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04233075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04457476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04401008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04401012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04400998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10520704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520699 - -
          Affymetrix HT MG-430 PM Array Plate 1437814_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10520698 - -
          Mouse Genome 430 2.0 Array 1437814_PM_at - -
          Mouse Genome 430 2.0 Array 1437814_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231098_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231098_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500002668.mm.1 TC0500000312.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002668.mm.1 NM_177677.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002668.mm.1 NM_177677.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002667.mm.1 TC0500000312.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002667.mm.1 NM_177677.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002667.mm.1 NM_177677.3 Refseq
          Mouse Expression Set 430 1437814_PM_at - -
          Mouse Expression Set 430 1437814_at - -
          Affymetrix HT Mouse Genome 430B Array 1437814_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437814_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 518261 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2030102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666340 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666340 - -
          mouse-6 v1.1 (Illumina) scl27962.6_713-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27962.6_713-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2666340 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0