Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400018140.mm.1 |
TC0400002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018140.mm.1 |
NM_145711.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018140.mm.1 |
NM_145711.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018141.mm.1 |
TC0400002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018141.mm.1 |
NM_145711.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018141.mm.1 |
NM_145711.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018137.mm.1 |
TC0400002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018137.mm.1 |
NM_145711.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018137.mm.1 |
NM_145711.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018142.mm.1 |
TC0400002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018142.mm.1 |
NM_145711.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018142.mm.1 |
NM_145711.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018143.mm.1 |
TC0400002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018143.mm.1 |
NM_145711.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018143.mm.1 |
NM_145711.4 |
Refseq |