Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289476.1 | NP_001276405.1 | 3 - 4 | 1077 | 63 | |
NM_011969.2 | NP_036099.1 | 3 - 4 | 438 | 63 | ||
GenBank mRNA | AF019662.1 | - | 3 - 4 | 376 | 63 | |
AK088615.1 | - | 3 - 4 | 383 | 63 | ||
AK146765.1 | - | 3 - 4 | 395 | 63 | ||
AK165563.1 | - | 3 - 4 | 391 | 63 | ||
AK168234.1 | - | 3 - 4 | 409 | 63 | ||
BC008222.1 | - | 3 - 4 | 371 | 2581897 | 63 | |
BC014752.1 | - | 3 - 4 | 1025 | 63 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |