Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020295.3 | NP_064691.2 | 9 - 10 | 956 | 129 | |
GenBank mRNA | AF190665.1 | - | 9 - 10 | 758 | 129 | |
AK030631.1 | - | 10 - 11 | 1762 | 129 | ||
AK195974.1 | - | 2 - 3 | 147 | 129 | ||
BC016110.1 | - | 9 - 10 | 715 | 4481088 | 129 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |