Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01227803_m1
          See other TUT1 GE Assays ›
          Gene Symbol:
          Tut1
          Gene Name:
          terminal uridylyl transferase 1, U6 snRNA-specific
          Gene Aliases:
          2700038E08Rik, PAPD2, Rbm21, TUTase6
          Chromosome Location:
          Chr.19: 8953850 - 8966210 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01227803_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tut1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_197993.2 NP_932110.1 5 - 6 1262 79
          GenBank mRNA BC023900.1 - 5 - 6 1239 5321265 79
          BC025499.1 - 5 - 6 1286 5320398 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000169.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tut1

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA polyadenylation
          mRNA cleavage
          mRNA processing
          snRNA processing
          nucleotide binding
          nucleic acid binding
          RNA binding
          mRNA 3'-UTR binding
          polynucleotide adenylyltransferase activity
          ATP binding
          transferase activity
          nucleotidyltransferase activity
          enzyme binding
          metal ion binding
          RNA uridylyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986199 BC023900.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986199 BC025499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986199 AK215597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986199 NM_197993.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126118 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173746 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986199 BC023900.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986199 BC025499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986199 AK215597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986199 NM_197993.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433556 BC025499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433556 NM_197993.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134136 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123892 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14418063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342729 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342734 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461355 - -
          Affymetrix HT MG-430 PM Array Plate 1451106_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440395_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461354 - -
          Mouse Genome 430 2.0 Array 1440395_at - -
          Mouse Genome 430 2.0 Array 1440395_PM_at - -
          Mouse Genome 430 2.0 Array 1451106_at - -
          Mouse Genome 430 2.0 Array 1451106_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70044_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70044_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002316.mm.1 TC1900000169.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002316.mm.1 NM_197993.3 Refseq
          Mouse Expression Set 430 1451106_at - -
          Mouse Expression Set 430 1451106_PM_at - -
          Mouse Expression Set 430 1440395_PM_at - -
          Mouse Expression Set 430 1440395_at - -
          Mouse Genome 430A 2.0 Array 1451106_PM_at - -
          Mouse Genome 430A 2.0 Array 1451106_at - -
          Affymetrix HT Mouse Genome 430A Array 1451106_at - -
          Affymetrix HT Mouse Genome 430A Array 1451106_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440395_at - -
          Affymetrix HT Mouse Genome 430B Array 1440395_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 302285 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6350309 - -
          Illumina mouse-6 v1.0 expression beadchip 105340671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232593 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232593 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215298 - -
          mouse-6 v1.1 (Illumina) scl53393.10_649-S - -
          Illumina Sentrix mouse-6.v1 bead array 105340671 - -
          Illumina Sentrix mouse-6.v1 bead array 6350309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53393.10_649-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105340671 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215298 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53393.10_649-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0