Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01227886_g1
          See other SLC22A20 GE Assays ›
          Gene Symbol:
          Slc22a20
          Gene Name:
          solute carrier family 22 (organic anion transporter), member 20
          Gene Aliases:
          Gm963, OAT1-L, mOAT6
          Chromosome Location:
          Chr.19: 5970232 - 5986551 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01227886_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198650.2 NP_941052.1 8 - 9 1373 86
          GenBank mRNA AY194118.1 - 8 - 9 1372 86
          BC046588.1 - 8 - 9 1373 6309674 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000994.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a20

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          organic anion transport
          transmembrane transport
          anion transmembrane transport
          organic anion transmembrane transporter activity
          transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081966 BC046588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081966 NM_198650.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75561 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176525 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128673 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081966 BC046588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081966 NM_198650.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P136249 BC046588.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P136249 AY194118.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P136249 NM_198650.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422578 AK053009.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932055 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176525 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128673 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09094999 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09095016 - -
          Whole Mouse Genome G4122A A_51_P422578 AK053009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136249 BC046588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136249 AY194118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136249 NM_198650.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122341 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176525 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14480528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14480519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14381090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14174044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14172950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14340041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14480524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14483523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14174042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14340036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14316905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14256934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14367896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14157970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14360678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465306 - -
          Affymetrix Mouse Gene 1.1 ST Array 10465303 - -
          Affymetrix Mouse Clariom S Array PSR1900009093.mm.1 TC1900000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009093.mm.1 NM_198650.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009093.mm.1 NM_198650.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009094.mm.1 TC1900000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009094.mm.1 NM_198650.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009094.mm.1 NM_198650.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358303 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1190746 - -
          Illumina mouse-6 v1.0 expression beadchip 1190746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190746 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2697153 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2697153 - -
          mouse-6 v1.1 (Illumina) scl0381203.8_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190746 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0381203.8_26-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0