Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01228833_m1
          See other 5530601H04RIK GE Assays ›
          Gene Symbol:
          5530601H04Rik
          Gene Name:
          RIKEN cDNA 5530601H04 gene
          Gene Aliases:
          AI835882
          Chromosome Location:
          Chr.X: 105040854 - 105070124 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01228833_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5530601H04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015467.1 3 - 4 350 70
          GenBank mRNA AK017459.1 - 287 70
          AK043766.1 - 323 70
          AK047017.1 - 323 70
          AK080296.1 - 3 - 4 318 70
          AK084653.1 - 3 - 4 323 70
          AK084980.1 - 3 - 4 302 70
          AK085098.1 - 3 - 4 295 70
          AK085485.1 - 3 - 4 289 70
          AK157237.1 - 281 70
          AK164650.1 - 3 - 4 351 70
          BC049927.1 - 139 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB660538.1 - 3 - 4 351 Chr. X - 105043908 - 105070124 [-] on Build GRCm38 70
          GenBankmRNA BB646114.1 - 3 - 4 323 Chr. X - 105039687 - 105070096 [-] on Build GRCm38 70
          GenBankmRNA BU614208.1 - 3 - 4 334 Chr. X - 105039580 - 105070095 [-] on Build GRCm38 70
          GenBankmRNA BY214823.1 - 3 - 4 278 Chr. X - 105066809 - 105070117 [-] on Build GRCm38 70
          GenBankmRNA AA881751.1 - 3 - 4 269 Chr. X - 105066813 - 105070108 [-] on Build GRCm38 70
          GenBankmRNA BX527550.1 - 3 - 4 270 Chr. X - 105044076 - 105070108 [-] on Build GRCm38 70
          GenBankmRNA BY214598.1 - 3 - 4 278 Chr. X - 105066797 - 105070117 [-] on Build GRCm38 70
          GenBankmRNA DV066789.1 - 3 - 4 370 Chr. X - 105043168 - 105070087 [-] on Build GRCm38 70
          GenBankmRNA CF749494.1 - 3 - 4 284 Chr. X - 105042749 - 105070058 [-] on Build GRCm38 70
          GenBankmRNA BY198982.1 - 3 - 4 287 Chr. X - 105066808 - 105070060 [-] on Build GRCm38 70
          GenBankmRNA CA886904.2 - 3 - 4 346 Chr. X - 105047909 - 105070107 [-] on Build GRCm38 70
          GenBankmRNA BE652703.1 - 1 - 2 63 Chr. X - 105044085 - 105068531 [-] on Build GRCm38 70
          GenBankmRNA BG695296.1 - 3 - 4 314 Chr. X - 105043978 - 105070085 [-] on Build GRCm38 70
          GenBankmRNA BB659196.1 - 3 - 4 323 Chr. X - 105044021 - 105070096 [-] on Build GRCm38 70
          GenBankmRNA BB661289.1 - 3 - 4 287 Chr. X - 105043986 - 105070060 [-] on Build GRCm38 70
          GenBankmRNA CK619918.1 - 3 - 4 273 Chr. X - 105044061 - 105070047 [-] on Build GRCm38 70
          GenBankmRNA BI691748.1 - 159 Chr. - - [-] on Build 70
          GenBankmRNA BY745619.1 - 3 - 4 318 Chr. X - 105044039 - 105070092 [-] on Build GRCm38 70
          GenBankmRNA BB660227.1 - 3 - 4 295 Chr. X - 105044021 - 105070068 [-] on Build GRCm38 70
          GenBankmRNA DV066765.1 - 3 - 4 365 Chr. X - 105039541 - 105070119 [-] on Build GRCm38 70
          GenBankmRNA BY018699.1 - 3 - 4 295 Chr. X - 105066777 - 105070068 [-] on Build GRCm38 70
          GenBankmRNA BG297259.1 - 294 Chr. - - [-] on Build 70
          GenBankmRNA BY723805.1 - 3 - 4 337 Chr. X - 105039667 - 105070111 [-] on Build GRCm38 70
          GenBankmRNA BB659892.1 - 3 - 4 302 Chr. X - 105044025 - 105070071 [-] on Build GRCm38 70
          GenBankmRNA BB641536.1 - 3 - 4 323 Chr. X - 105039797 - 105070096 [-] on Build GRCm38 70
          GenBankmRNA BF661519.1 - 3 - 4 285 Chr. X - 105043983 - 105070059 [-] on Build GRCm38 70
          GenBankmRNA BE647238.1 - 3 - 4 283 Chr. X - 105044019 - 105070057 [-] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream melanoma antigen,
          family E,
          2
          Magee2 9630059J11Rik,
          BB131182,
          Mage-e2
          -209.5kb Sense Chr. X - 104854952 - 104857267 on NCBI Build GRCm38
          Downstream polysaccharide biosynthesis domain containing 1 Pbdc1 2610029G23Rik 11.1kb Anti-sense Chr. X - 105079756 - 105117094 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002807.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085460 BC055463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085460 AK164650.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085460 AK085485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085460 AK084980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085460 AK085098.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 AK036629.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 AK049737.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 AY024346.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 AY238936.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 BC019366.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 BC029774.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 BC031846.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 BC059816.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 BC080810.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632193 NM_153055.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120679 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149946 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085460 BC055463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085460 AK164650.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085460 AK085485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085460 AK084980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085460 AK085098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09383025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09383014 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09383055 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149946 - -
          Whole Mouse Genome G4122A A_51_P172355 AK164650.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172355 AK084980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172355 AK085098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172355 AK084653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172355 NR_015467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145878 BC049927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145878 AK157237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145878 AK164650.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145878 AK085485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145878 AK084980.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124459 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14915932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14915925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15009806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15059377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14915926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14704322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14915937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15009799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14743834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15059375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14717966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578978 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1436347_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1438893_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452750_PM_at - -
          Mouse Genome 430 2.0 Array 1436347_a_at - -
          Mouse Genome 430 2.0 Array 1452750_PM_at - -
          Mouse Genome 430 2.0 Array 1438893_PM_at - -
          Mouse Genome 430 2.0 Array 1452750_at - -
          Mouse Genome 430 2.0 Array 1436347_PM_a_at - -
          Mouse Genome 430 2.0 Array 1438893_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71445_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71445_at - -
          Affymetrix Mouse Clariom S Array PSR0X00017693.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017693.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017693.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017691.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017691.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017691.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017692.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017692.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017692.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017716.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017716.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017716.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017717.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017717.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017717.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017690.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017690.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017690.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017694.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017694.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017694.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017696.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017696.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017696.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017697.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017697.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017697.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017698.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017698.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017698.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017699.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017699.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017699.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017700.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017700.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017700.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017705.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017705.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017705.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017721.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017721.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017721.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017689.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017689.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017689.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017704.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017704.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017704.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017713.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017713.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017713.mm.1 NR_015467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017724.mm.1 TC0X00002807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017724.mm.1 NR_015467.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017724.mm.1 NR_015467.1 Refseq
          Mouse Expression Set 430 1438893_at - -
          Mouse Expression Set 430 1452750_PM_at - -
          Mouse Expression Set 430 1452750_at - -
          Mouse Expression Set 430 1436347_PM_a_at - -
          Mouse Expression Set 430 1436347_a_at - -
          Mouse Expression Set 430 1438893_PM_at - -
          Murine Genome U74v2 Set 166905_at - -
          Murine Genome U74v2 Set 166905_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452750_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436347_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1452750_at - -
          Affymetrix HT Mouse Genome 430B Array 1438893_at - -
          Affymetrix HT Mouse Genome 430B Array 1438893_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436347_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101190279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0