Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01229475_g1
          See other GM3942 GE Assays ›
          Gene Symbol:
          Gm3942
          Gene Name:
          predicted gene 3942
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 112837733 - 112856525 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01229475_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm3942
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK036450.1 - 3 - 4 480 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700010J16 gene 1700010J16Rik -56.6kb Sense Chr. 10 - 112726926 - 112785599 on NCBI Build GRCm38
          Downstream ataxin 7-like 3B Atxn7l3b 4921506J03Rik,
          6230409E21Rik,
          AI315132,
          ENSMUSG00000074747
          83.0kb Anti-sense Chr. 10 - 112925428 - 112929026 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125770 AK036450.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386438 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125770 AK036450.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125770 AK036450.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08332628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08525682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08614981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08708157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08579073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08651049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08708154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08651048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27456182 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0