Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01229641_g1
          See other E330032C10RIK GE Assays ›
          Gene Symbol:
          E330032C10Rik
          Gene Name:
          RIKEN cDNA E330032C10 gene
          Gene Aliases:
          E330032C10
          Chromosome Location:
          Chr.17: 73250986 - 73260771 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01229641_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E330032C10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK143670.1 - 3 - 4 2027 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB736367.1 - 3 - 4 Chr. 17 - 73252628 - 73254146 [+] on Build GRCm38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream lysocardiolipin acyltransferase 1 Lclat1 AI181996,
          Agpat8,
          Alcat1,
          Gm91,
          Lycat
          -10.4kb Sense Chr. 17 - 73107985 - 73243368 on NCBI Build GRCm38
          Downstream calpain 13 Capn13 CANP 13,
          Gm943
          52.3kb Anti-sense Chr. 17 - 73306456 - 73399306 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2258159 AK054493.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18663 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03366611 - -
          Whole Mouse Genome G4122A A_52_P1076168 AK054493.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13151873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13151870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13151869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13432056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31345061 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline