Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133984.2 | NP_598745.2 | 8 - 9 | 993 | 64 | |
GenBank mRNA | AK088071.1 | - | 7 - 8 | 1298 | 64 | |
BC011431.1 | - | 7 - 8 | 938 | 3981356 | 64 | |
BC024428.1 | - | 7 - 8 | 798 | 4971671 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |