Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01230973_g1
          See other 1810009A15RIK GE Assays ›
          Gene Symbol:
          1810009A15Rik Lbhd1
          Gene Name:
          RIKEN cDNA 1810009A15 gene
          LBH domain containing 1
          Gene Aliases:
          -
          Gm21743
          Chromosome Location:
          Chr.19: 8888883 - 8890759 on Build GRCm38
          Chromosome Location:
          Chr.19: 8883878 - 8890759 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01230973_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810009A15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025463.3 NP_079739.1 3 - 4 309 73
          NR_104416.1 2 - 3 468 73
          GenBank mRNA AK007390.1 - 3 - 4 290 73
          AK012061.1 - 3 - 4 299 73
          BC047099.1 - 3 - 4 288 5253822 73
          BC058702.1 - 3 - 4 272 6822952 73
          Gene Symbol: Lbhd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286103.1 NP_001273032.1 5 - 6 805 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000163.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1810009A15Rik
          Click here to find siRNAs for Gene Lbhd1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968035 BC047099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968035 BC058702.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676510 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119560 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968035 BC047099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968035 BC058702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126647 BC047099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126647 BC058702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126647 NM_025463.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126647 AK007390.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103213 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467079 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14479067 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14479068 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467076 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14289592 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467080 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461271 - -
          Affymetrix HT MG-430 PM Array Plate 1417886_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461268 - -
          Mouse Genome 430 2.0 Array 1417886_PM_at - -
          Mouse Genome 430 2.0 Array 1417886_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66276_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66276_at - -
          Affymetrix Mouse Clariom S Array PSR1900002193.mm.1 TC1900000163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002193.mm.1 NM_001286103.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002193.mm.1 NM_025463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002193.mm.1 NR_104416.1 Refseq
          Mouse Expression Set 430 1417886_at - -
          Mouse Expression Set 430 1417886_PM_at - -
          Murine Genome U74v2 Set 103565_at - -
          Murine Genome U74v2 Set 103565_PM_at - -
          Mouse Genome 430A 2.0 Array 1417886_at - -
          Mouse Genome 430A 2.0 Array 1417886_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 307146 - -
          ABI Mouse Genome Survey Microarray 899524 - -
          ABI Mouse Genome Survey Microarray 903347 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5360133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221262 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221262 - -
          mouse-6 v1.1 (Illumina) scl066276.2_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066276.2_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2872246 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221262 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221262 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2872246 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066276.2_21-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-x7z22:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0