Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053105.2 | NP_444335.2 | 4 - 5 | 1737 | 129 | |
GenBank mRNA | AK031932.1 | - | 4 - 5 | 1549 | 129 | |
AK220397.1 | - | 4 - 5 | 1041 | 129 | ||
BC094584.1 | - | 4 - 5 | 1737 | 30547502 | 129 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |