Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2362955 |
AK077069.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
185350 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346622 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
145520 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304848 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
830763 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
263257 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
AB035123.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
AB035174.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
AK030648.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
AK035447.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
AK157562.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
AK159158.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
BC022924.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
BC036985.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
NM_001025310.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
NM_001025311.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385469 |
NM_016973.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
182837 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
883306 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
873024 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
285873 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
275170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553964 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
260629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547007 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346508 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
46081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
303422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689045 |
AK006590.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
4055 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
620610 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
352995 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
158219 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
373847 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554300 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
59792 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
100399 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2191 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
131005 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
46861 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P956292 |
AK077069.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P111057 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07980049 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728265 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728260 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728266 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728259 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728251 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728250 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728256 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07721058 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728252 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728248 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728262 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26728264 |
- |
- |