Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159529.1 | NP_001153001.1 | 1 - 2 | 132 | 82 | |
NM_009187.3 | NP_033213.1 | 1 - 2 | 132 | 82 | ||
GenBank mRNA | AK133520.1 | - | 1 - 2 | 132 | 82 | |
AK150034.1 | - | 1 - 2 | 129 | 82 | ||
AK152553.1 | - | 1 - 2 | 122 | 82 | ||
AK159441.1 | - | 1 - 2 | 130 | 82 | ||
AK166761.1 | - | 1 - 2 | 117 | 82 | ||
AK167062.1 | - | 1 - 2 | 127 | 82 | ||
BC004715.1 | - | 1 - 2 | 96 | 3582142 | 82 | |
BC085296.1 | - | 1 - 2 | 101 | 5337317 | 82 | |
BY122624.1 | - | 1 - 2 | 132 | 82 | ||
X80899.1 | - | 1 - 2 | 100 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |