Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011500.2 | NP_035630.2 | 4 - 5 | 564 | 108 | |
GenBank mRNA | AJ223777.1 | - | 4 - 5 | 491 | 108 | |
AK051549.1 | - | 4 - 5 | 566 | 108 | ||
AK136032.1 | - | 2 - 3 | 213 | 108 | ||
BC090968.1 | - | 4 - 5 | 564 | 30646517 | 108 | |
BC150727.1 | - | 4 - 5 | 532 | 9087638 | 108 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |