Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
TC0900003009.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
NM_001110271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
NM_145919.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
NM_001110271.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
NM_001110272.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
NM_001110272.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024994.mm.1 |
NM_145919.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
TC0900003009.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
NM_001110271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
NM_001110271.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
NM_001110272.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
NM_001110272.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
NM_145919.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024992.mm.1 |
NM_145919.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024998.mm.1 |
TC0900003009.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024998.mm.1 |
NM_001110271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024998.mm.1 |
NM_001110271.1 |
Refseq |