Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01233887_m1
          See other LMBRD1 GE Assays ›
          Gene Symbol:
          Lmbrd1
          Gene Name:
          LMBR1 domain containing 1
          Gene Aliases:
          0910001K20Rik, AV347960
          Chromosome Location:
          Chr.1: 24678544 - 24766301 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01233887_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lmbrd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026719.2 NP_080995.2 2 - 3 448 103
          GenBank mRNA AK152170.1 - 2 - 3 259 103
          AK153237.1 - 2 - 3 259 103
          AK158982.1 - 2 - 3 284 103
          AK167080.1 - 2 - 3 267 103
          AY061989.1 - 1 - 2 159 103
          BC025579.1 - 2 - 3 280 5325113 103
          BC027774.1 - 2 - 3 331 5346846 103
          BC031902.1 - 2 - 3 271 5362166 103
          BC039053.1 - 2 - 3 362 2655321 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000171.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK158982.1, AY061989.1, BC025579.1, BC027774.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lmbrd1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          insulin receptor internalization
          regulation of MAPK cascade
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          negative regulation of protein kinase B signaling
          insulin receptor binding
          cobalamin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100001092.mm.1 TC0100000171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001092.mm.1 NM_026719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001092.mm.1 NM_026719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001094.mm.1 TC0100000171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001094.mm.1 NM_026719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001094.mm.1 NM_026719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001090.mm.1 TC0100000171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001090.mm.1 NM_026719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001090.mm.1 NM_026719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001096.mm.1 TC0100000171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001096.mm.1 NM_026719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001096.mm.1 NM_026719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001091.mm.1 TC0100000171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001091.mm.1 NM_026719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001091.mm.1 NM_026719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001097.mm.1 TC0100000171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001097.mm.1 NM_026719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001097.mm.1 NM_026719.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 381010 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0