Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01234782_m1
          See other B930071A02 GE Assays ›
          Gene Symbol:
          B930071A02
          Gene Name:
          uncharacterized protein B930071A02
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01234782_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B930071A02
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK081028.1 - 2 - 3 581 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY727984.1 - 2 - 3 581 Chr. 10 - 94570083 - 94612085 [-] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 4792 Gm4792 E130012K09,
          EG215472
          -269.1kb Sense Chr. 10 - 94293663 - 94298703 on NCBI Build GRCm38
          Downstream microRNA 7211 Mir7211 Gm27536,
          mmu-mir-7211
          13.1kb Sense Chr. 10 - 94586137 - 94586199 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774026 AK004781.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774026 AK076707.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774026 D49473.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774026 D49474.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774026 NM_011441.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558764 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07156342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02192440 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127719 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07156342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08677636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08445081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08701008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08445089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08861681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08790501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08816041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08445091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08677634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08907981 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 333489 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0