Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01234880_g1
          See other GM614 GE Assays ›
          Gene Symbol:
          Gm614
          Gene Name:
          predicted gene 614
          Gene Aliases:
          Il2rg
          Chromosome Location:
          Chr.X: 101261377 - 101263992 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01234880_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm614
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033362.2 NP_001028534.1 5 - 6 619 72
          GenBank mRNA AK131750.1 - 5 - 6 620 72
          BC048573.1 - 5 - 6 629 72
          BC147523.1 - 5 - 6 616 9056515 72
          BC147537.1 - 5 - 6 616 9056528 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003436.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012216 BC048573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012216 BC147523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012216 BC147537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110623 NM_001033362.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110623 AK131750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110623 BC048573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110623 BC147523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110623 BC147537.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118096 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110623 NM_001033362.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110623 AK131750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110623 BC048573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110623 BC147523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110623 BC147537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012216 BC048573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012216 BC147523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012216 BC147537.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09375361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32563141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14934377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14546992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15003895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15003904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32563126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606011 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606009 - -
          Mouse Genome 430 2.0 Array 1458481_PM_at - -
          Mouse Genome 430 2.0 Array 1458481_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 245536_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 245536_at - -
          Affymetrix Mouse Clariom S Array PSR0X00021173.mm.1 TC0X00003436.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00021173.mm.1 NM_001033362.3 Refseq
          Mouse Expression Set 430 1458481_at - -
          Mouse Expression Set 430 1458481_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458481_at - -
          Affymetrix HT Mouse Genome 430B Array 1458481_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 677420 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2784703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2784703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0