Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235176_m1
          See other 4933430M04RIK GE Assays ›
          Gene Symbol:
          4933430M04Rik
          Gene Name:
          RIKEN cDNA 4933430M04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 24481909 - 24569951 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235176_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933430M04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045857.2 3 - 4 295 148
          GenBank mRNA AK016999.1 - 3 - 4 296 148
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY716684.1 - 3 - 4 296 Chr. 11 - 24481909 - 24569952 [+] on Build GRCm38 148

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream B cell CLL/lymphoma 11A (zinc finger protein) Bcl11a 2810047E18Rik,
          BCL-11A,
          Ctip1,
          D930021L15Rik,
          Evi9,
          Evi9a,
          Evi9b,
          Evi9c,
          mKIAA1809
          -386.9kb Sense Chr. 11 - 24076564 - 24173558 on NCBI Build GRCm38
          Downstream predicted gene 10466 Gm10466 ENSMUSG00000073103 151.1kb Anti-sense Chr. 11 - 24721006 - 24730722 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000258.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1400001161.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97841 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07281212 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127579 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07281212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09425703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09309717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09433788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09433776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09503067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09575033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09496931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09425706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09454068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09291396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09454064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09425709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09642179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09309734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09291408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09672496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09323660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09323662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09425712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09532079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09345643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09288027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09496926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09433785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09327795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27652042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09623644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27651763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09288023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09454066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09553905 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100002222.mm.1 TC1100000258.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100002222.mm.1 NR_045857.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 527527 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104610440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0