Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236149_g1
          See other NAA40 GE Assays ›
          Gene Symbol:
          Naa40
          Gene Name:
          N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae)
          Gene Aliases:
          4931433E08Rik, AU023197, Nat11
          Chromosome Location:
          Chr.19: 7225668 - 7241222 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236149_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Naa40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027643.1 NP_081919.1 7 - 8 855 69
          GenBank mRNA AK033510.1 - 7 - 8 677 69
          AK033896.1 - 7 - 8 686 69
          AK035418.1 - 6 - 7 1573 69
          AK170548.1 - 7 - 8 681 69
          AK202080.1 - 2 - 3 136 69
          AK205460.1 - 1 - 2 88 69
          AK209446.1 - 2 - 3 135 69
          BC020020.1 - 7 - 8 706 3156924 69
          BC049808.1 - 1 - 2 93 69
          BC058212.1 - 2 - 3 514 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001037.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          molecular_function
          dihydrolipoamide branched chain acyltransferase activity
          sterol O-acyltransferase activity
          N-acetyltransferase activity
          O-acyltransferase activity
          palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
          carnitine O-acyltransferase activity
          acetyltransferase activity
          C-acyltransferase activity
          palmitoyltransferase activity
          N-acyltransferase activity
          acylglycerol O-acyltransferase activity
          serine O-acyltransferase activity
          O-acetyltransferase activity
          O-octanoyltransferase activity
          octanoyltransferase activity
          O-palmitoyltransferase activity
          S-acyltransferase activity
          S-acetyltransferase activity
          S-malonyltransferase activity
          malonyltransferase activity
          C-palmitoyltransferase activity
          transferase activity
          acyltransferase activity
          succinyltransferase activity
          N-succinyltransferase activity
          O-succinyltransferase activity
          S-succinyltransferase activity
          sinapoyltransferase activity
          O-sinapoyltransferase activity
          peptidyl-lysine N6-myristoyltransferase activity
          peptidyl-lysine N6-palmitoyltransferase activity
          benzoyl acetate-CoA thiolase activity
          3-hydroxybutyryl-CoA thiolase activity
          3-ketopimelyl-CoA thiolase activity
          N-palmitoyltransferase activity
          myristoyltransferase activity
          acyl-CoA N-acyltransferase activity
          protein-cysteine S-myristoyltransferase activity
          protein-cysteine S-acyltransferase activity
          dihydrolipoamide S-acyltransferase activity
          glucosaminyl-phosphatidylinositol O-acyltransferase activity
          ergosterol O-acyltransferase activity
          lanosterol O-acyltransferase activity
          naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
          2-methylhexanoyl-CoA C-acetyltransferase activity
          butyryl-CoA 2-C-propionyltransferase activity
          2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
          alpha-aminoadipate acetyltransferase activity
          keto acid formate lyase activity
          Ras palmitoyltransferase activity
          azetidine-2-carboxylic acid acetyltransferase activity
          peptidyl-lysine acetyltransferase activity
          acetyl-CoA:L-lysine N6-acetyltransferase
          UDP-3-O-(R-3-hydroxymyristoyl)-glucosamine N-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155062 BC058212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155062 BC020020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155062 BC049808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155062 BB112808.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155062 AK199921.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786441 AY042193.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786441 NM_153067.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143332 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157010 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175609 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155062 BC058212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155062 BC020020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155062 BC049808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155062 BB112808.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155062 AK199921.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09098241 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175609 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157010 - -
          Whole Mouse Genome G4122A A_52_P684579 BC058212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P684579 BC020020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P684579 BC049808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P684579 AK217157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P684579 AK181058.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175609 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14144298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14213174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14148301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14184387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14255689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14360488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14175995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14143676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14247486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14415260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14226015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14420735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14164967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14184385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14247485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14457954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14255684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14164971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14315524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14315530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31938767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465647 - -
          Affymetrix HT MG-430 PM Array Plate 1452120_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443625_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458576_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465638 - -
          Mouse Genome 430 2.0 Array 1452120_at - -
          Mouse Genome 430 2.0 Array 1443625_at - -
          Mouse Genome 430 2.0 Array 1443625_PM_at - -
          Mouse Genome 430 2.0 Array 1452120_PM_at - -
          Mouse Genome 430 2.0 Array 1458576_PM_at - -
          Mouse Genome 430 2.0 Array 1458576_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70999_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70999_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900009676.mm.1 TC1900001037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009676.mm.1 NM_027643.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009676.mm.1 NM_027643.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009677.mm.1 TC1900001037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009677.mm.1 NM_027643.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009677.mm.1 NM_027643.1 Refseq
          Mouse Expression Set 430 1452120_at - -
          Mouse Expression Set 430 1452120_PM_at - -
          Mouse Expression Set 430 1443625_PM_at - -
          Mouse Expression Set 430 1443625_at - -
          Mouse Expression Set 430 1458576_PM_at - -
          Mouse Expression Set 430 1458576_at - -
          Murine Genome U74v2 Set 160191_PM_at - -
          Murine Genome U74v2 Set 160191_at - -
          Mouse Genome 430A 2.0 Array 1452120_PM_at - -
          Mouse Genome 430A 2.0 Array 1452120_at - -
          Affymetrix HT Mouse Genome 430A Array 1452120_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452120_at - -
          Affymetrix HT Mouse Genome 430B Array 1443625_at - -
          Affymetrix HT Mouse Genome 430B Array 1458576_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458576_at - -
          Affymetrix HT Mouse Genome 430B Array 1443625_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101090463 - -
          Illumina mouse-6 v1.0 expression beadchip 101090463 - -
          Illumina mouse-6 v1.0 expression beadchip 100730504 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2531537 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228701 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256960 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2558125 - -
          Illumina Sentrix mouse-6.v1 bead array 101090463 - -
          Illumina Sentrix mouse-6.v1 bead array 100730504 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101090463 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100730504 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2545383 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2558125 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0