Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01237743_s1
          See other ZRSR1 GE Assays ›
          Gene Symbol:
          Zrsr1
          Gene Name:
          zinc finger (CCCH type), RNA binding motif and serine/arginine rich 1
          Gene Aliases:
          D11Ncvs75, Irlgs2, SP2, U2af1-rs1, U2afbp-rs
          Chromosome Location:
          Chr.11: 22972005 - 22976496 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01237743_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zrsr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011663.3 NP_035793.1 1 2694 75
          GenBank mRNA AK030552.1 - 1 1484 75
          D17407.1 - 2 2639 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000231.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zrsr1

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          nucleic acid binding
          RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643661 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643660 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643663 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643662 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643659 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 D17407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 S69507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 NM_011663.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 AK030552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 D17407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 S69507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 NM_011663.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407675 AK030552.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102717 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 D17407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 S69507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 NM_011663.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 AK030552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 D17407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 S69507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 NM_011663.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407675 AK030552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 D17407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 S69507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 NM_011663.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 AK030552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 D17407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 S69507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 NM_011663.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407675 AK030552.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09127883 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09127885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09127884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643663 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643659 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643661 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643662 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27643660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27643674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09127894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09670934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09366035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09362730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09362727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09362726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09127885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09620743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09362731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27643633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09620738 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10384666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374561 - -
          Affymetrix HT MG-430 PM Array Plate 1449354_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10374560 - -
          Affymetrix Mouse Gene 1.1 ST Array 10384662 - -
          Mouse Genome 430 2.0 Array 1449354_PM_at - -
          Mouse Genome 430 2.0 Array 1449354_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22183_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22183_at - -
          Affymetrix Mouse Clariom S Array PSR1100001862.mm.1 TC1100000231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100001862.mm.1 NM_011663.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001862.mm.1 NM_011663.3 Refseq
          Mouse Expression Set 430 1449354_at - -
          Mouse Expression Set 430 1449354_PM_at - -
          Murine Genome U74v2 Set 102817_PM_at - -
          Murine Genome U74v2 Set 102817_at - -
          Mouse Genome 430A 2.0 Array 1449354_at - -
          Mouse Genome 430A 2.0 Array 1449354_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449354_at - -
          Affymetrix HT Mouse Genome 430A Array 1449354_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 730858 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4010463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2448615 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2448615 - -
          mouse-6 v1.1 (Illumina) scl022183.2_94-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022183.2_94-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2448615 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2448615 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0