Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003911.2 | NP_001003911.2 | 22 - 23 | 4791 | 73 | |
GenBank mRNA | AK052239.1 | - | 1 - 2 | 947 | 73 | |
AY551090.1 | - | 23 - 24 | 4602 | 73 | ||
BC030442.1 | - | 10 - 11 | 1232 | 73 | ||
BC058991.1 | - | 19 - 20 | 3763 | 73 | ||
BC076575.1 | - | 21 - 22 | 4499 | 73 | ||
BC141173.1 | - | 22 - 23 | 4567 | 9055785 | 73 | |
BC145491.1 | - | 23 - 24 | 4390 | 9054054 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |