Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01240082_m1
          See other 2900079G21RIK GE Assays ›
          Gene Symbol:
          2900079G21Rik
          Gene Name:
          RIKEN cDNA 2900079G21 gene
          Gene Aliases:
          C130083B14, Gm6179
          Chromosome Location:
          Chr.9: 112234277 - 112348925 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01240082_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2900079G21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033431.1 1 - 2 104 64
          GenBank mRNA AK013806.1 - 1 - 2 105 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001493.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 BC056188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 BE946996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 AK038201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 AK013806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 NR_015468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 BC056188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 BE946996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 AK038201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 AK013806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P456243 NR_015468.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16475 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107152 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 BC056188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 BE946996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 AK038201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 AK013806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 NR_015468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 BC056188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 BE946996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 AK038201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 AK013806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P456243 NR_015468.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02032440 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107152 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02032338 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06929835 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02032401 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02032279 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06929925 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06930031 - -
          Whole Mouse Genome G4122A A_52_P41074 AK048592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P41074 NR_033432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 BC056188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 BE946996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 AK038201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 AK013806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 NR_015468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 BC056188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 BE946996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 AK038201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 AK013806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P456243 NR_015468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161637 AK047318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161637 NR_033433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134123 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107152 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02032401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07772797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07670962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07549826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08065465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07772811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07687710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07772813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07670965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07859919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08161026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07759893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07859902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07859913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08064184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07859916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07859900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07670950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07967336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08153643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07967333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07772816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26834372 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453323_PM_at - -
          Mouse Genome 430 2.0 Array 1453323_at - -
          Mouse Genome 430 2.0 Array 1453323_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900012521.mm.1 TC0900001493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900012521.mm.1 NR_015468.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012521.mm.1 NR_015468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012521.mm.1 NR_033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012521.mm.1 NR_033431.1 Refseq
          Mouse Expression Set 430 1453323_at - -
          Mouse Expression Set 430 1453323_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550532 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0