Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01240874_g1
          See other ADAD2 GE Assays ›
          Gene Symbol:
          Adad2
          Gene Name:
          adenosine deaminase domain containing 2
          Gene Aliases:
          4930403J07Rik
          Chromosome Location:
          Chr.8: 119612710 - 119616926 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01240874_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029428.1 NP_083704.1 7 - 8 1252 79
          GenBank mRNA AK015063.1 - 7 - 8 1252 79
          BC137980.1 - 7 - 8 1239 8860999 79

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA processing
          biological_process
          RNA binding
          adenosine deaminase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140042 BC137980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140042 NM_029428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140042 AK015063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140042 BC137980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140042 NM_029428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140042 AK015063.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 AK045954.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 AK047856.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 AK051332.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 AK159687.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 BC010675.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 NM_009255.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554463 X70296.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109567 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140042 BC137980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140042 NM_029428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140042 AK015063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140042 BC137980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140042 NM_029428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140042 AK015063.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06662092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109567 - -
          Whole Mouse Genome G4122A A_51_P140042 BC137980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140042 NM_029428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140042 AK015063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140042 BC137980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140042 NM_029428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140042 AK015063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305987 BC137980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305987 NM_029428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305987 AK015063.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140004 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10575911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575910 - -
          Affymetrix HT MG-430 PM Array Plate 1431898_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10575909 - -
          Mouse Genome 430 2.0 Array 1431898_PM_at - -
          Mouse Genome 430 2.0 Array 1431898_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75773_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75773_at - -
          Mouse Expression Set 430 1431898_at - -
          Mouse Expression Set 430 1431898_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431898_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431898_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 694457 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217544 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline